Propionic acid, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: Propanoic acid
- CAS Number:79-09-4
- Formula:C₃H₆O₂
- MDL Number:MFCD00002756
- Molecular Weight:74.08 g/mol
- Merck Index:13,07917
- Boiling Pt:141 °C (1013 hPa)
- Melting Pt:–21 °C
- Density:0.99 g/cm³ (20 °C)
- Flash Pt:49 °C
- UN:3463
- ADR:8,II
- :M-Clarity™ quality level = MQ300
- Storage Temperature:Ambient
- EINECS:201-176-3
- REACH:01-2119486971-24
3 Options Available Below
Product Details & Documents
Synonyms: Propanoic acid
- CAS Number:79-09-4
- Formula:C₃H₆O₂
- MDL Number:MFCD00002756
- Molecular Weight:74.08 g/mol
- Merck Index:13,07917
- Boiling Pt:141 °C (1013 hPa)
- Melting Pt:–21 °C
- Density:0.99 g/cm³ (20 °C)
- Flash Pt:49 °C
- UN:3463
- ADR:8,II
- :M-Clarity™ quality level = MQ300
- Storage Temperature:Ambient
- EINECS:201-176-3
- REACH:01-2119486971-24



