Order Entry
Export
Orders LinkContactUsLinkComponent
Propionic acid, Sigma-Aldrich®

Propionic acid, Sigma-Aldrich®

Corrosive
Irritant
Supplier:   Merck
Propionic acid, Sigma-Aldrich®
Supplier:  Merck
Certificates
Propionic acid, Sigma-Aldrich®

3 Options Available Below

Product Details & Documents
Synonyms:  Propanoic acid
  • CAS Number:79-09-4
  • Formula:C₃H₆O₂
  • MDL Number:MFCD00002756
  • Molecular Weight:74.08 g/mol
  • Merck Index:13,07917
  • Boiling Pt:141 °C (1013 hPa)
  • Melting Pt:–21 °C
  • Density:0.99 g/cm³ (20 °C)
  • Flash Pt:49 °C
  • UN:3463
  • ADR:8,II
  •  :M-Clarity™ quality level = MQ300
  • Storage Temperature:Ambient
  • EINECS:201-176-3
  • REACH:01-2119486971-24
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 67,00 €
    67,00 €
    1 * 1 kg
       67,00 €

  • 10 kg
    Log in for availability
    Log in for availability
    1 * 10 kg
    1 * 10 kg - 244,00 €
    244,00 €
    1 * 10 kg
       244,00 €

  • 25 kg
    Log in for availability
    Log in for availability
    1 * 25 kg
    1 * 25 kg - 365,00 €
    365,00 €
    1 * 25 kg
       365,00 €