Acetic acid 99 - 100% 99-100%
Supplier: Roth Carl
Certificates
About this item
Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
CAS Number:64-19-7
:
Formula:H₃CCOOH
MDL Number:MFCD00036152
Boiling Pt:118
Molecular Weight:60.05 g/mol
Melting Pt:17
Flash Pt:38.5
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8 II
:GHS02
:GHS05
Specifications
- Environmentally Preferable: