Sebacoyl dichloride
Glass bottle, 50 mL
Catalog # ROTH4472.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Sebacoyl chloride
- CAS Number:111-19-3
- Formula:C₁₀H₁₆Cl₂O₂
- MDL Number:MFCD00000770
- Molecular Weight:239.14 g/mol
- Boiling Pt:220 °C (1013 hPa)
- Melting Pt:–5 °C
- Density:1.12 g/cm³ (20 °C)
- Flash Pt:165 °C
- Storage Temperature:Ambient
- EINECS:203-843-4
- UN:3265
- ADR:8,II
Specifications
Specifications
- Pack type:Glass bottle
- Pk:50 mL



