Sebacoyl dichloride ≥97%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 97% CAS No.: 111-19-3 Molecular Formula: ClCO(CH2)8COCl Formula Weight: 239.14 Flash Point: 165°(329°F) Refractive Index: 1.4680 Odour: Acrid Melting Point: -5° Boiling Point: 161°/14mm Beilstein Reference: 1365665 UN No.: 2922 MDL No.: MFCD00000770
Synonyms: Sebacoyl chloride
- CAS Number:111-19-3
- Formula:C₁₀H₁₆Cl₂O₂
- MDL Number:MFCD00000770
- Molecular Weight:239.14 g/mol
- Boiling Pt:161 °C (14 mmHg)
- Melting Pt:–5 °C
- Density:1.121
- Flash Pt:165 °C (329 °F)
- Storage Temperature:Ambient
- EINECS:203-843-4
- UN:2922
- ADR:8,II
3 Options Available Below
Product Details & Documents
Purity: 97% CAS No.: 111-19-3 Molecular Formula: ClCO(CH2)8COCl Formula Weight: 239.14 Flash Point: 165°(329°F) Refractive Index: 1.4680 Odour: Acrid Melting Point: -5° Boiling Point: 161°/14mm Beilstein Reference: 1365665 UN No.: 2922 MDL No.: MFCD00000770
Synonyms: Sebacoyl chloride
- CAS Number:111-19-3
- Formula:C₁₀H₁₆Cl₂O₂
- MDL Number:MFCD00000770
- Molecular Weight:239.14 g/mol
- Boiling Pt:161 °C (14 mmHg)
- Melting Pt:–5 °C
- Density:1.121
- Flash Pt:165 °C (329 °F)
- Storage Temperature:Ambient
- EINECS:203-843-4
- UN:2922
- ADR:8,II



