Triethyl citrate
Supplier: Merck
Jump to:
About this item
Synonyms: Triethyl 2-hydroxypropane-1,2,3-tricarboxylate, Hydroxytricarballylic acid triethyl ester, Citric acid triethyl ester
- CAS Number:77-93-0
- Formula:C₁₂H₂₀O₇
- MDL Number:MFCD00009201
- Molecular Weight:276.29 g/mol
- Boiling Pt:294 °C (1013 hPa)
- Melting Pt:–55 °C
- Density:1.135 g/cm³ (25 °C)
- Flash Pt:150 °C
- EINECS:201-070-7
2 Options Available Below
Product Details & Documents
Synonyms: Triethyl 2-hydroxypropane-1,2,3-tricarboxylate, Hydroxytricarballylic acid triethyl ester, Citric acid triethyl ester
- CAS Number:77-93-0
- Formula:C₁₂H₂₀O₇
- MDL Number:MFCD00009201
- Molecular Weight:276.29 g/mol
- Boiling Pt:294 °C (1013 hPa)
- Melting Pt:–55 °C
- Density:1.135 g/cm³ (25 °C)
- Flash Pt:150 °C
- EINECS:201-070-7



