Methyl myristate
Catalog # ROTH3753.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 mL
Specifications
About this item
:
:242.4 g/mol
Boiling Pt:323 °C (1013 hPa)
Melting Pt:18 °C
Density:0.86715 g/cm³ (20 °C)
Flash Pt:140 °C
Storage Temperature:Refrigerator
EINECS:204-680-1
CAS Number:124-10-7
Formula:C₁₅H₃₀O₂
MDL Number:MFCD00008983