Order Entry
Export
Orders LinkContactUsLinkComponent
 

Methyl myristate ≥99%

Methyl myristate ≥99%
Certificates
Methyl myristate ≥99%
Methyl myristate ≥99%

2 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 124-10-7 Molecular Formula: C15H30O2 Formula Weight: 242.40 Flash Point: 112°(234°F) Refractive Index: 1.4562 Melting Point: 18° Boiling Point: 323° MDL No.: MFCD00008983

Synonyms:  Tetradecylic acid methyl ester, Myristic acid methyl ester, Tetradecanoic acid methyl ester, Methyl tetradecanoate
  • CAS Number:124-10-7
  • Formula:C₁₅H₃₀O₂
  • MDL Number:MFCD00008983
  • Molecular Weight:242.4 g/mol
  • Boiling Pt:323 °C
  • Melting Pt:18 °C
  • Density:0.855
  • Flash Pt:>112 °C (234 °F)
  • Storage Temperature:Ambient
  • EINECS:204-680-1
Product Family Options

Product Information