Ethyl formate
Glass bottle, 1 L
Catalog # ROTH3574.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Formic acid ethyl ester
- CAS Number:109-94-4
- Formula:C₃H₆O₂
- MDL Number:MFCD00003294
- Molecular Weight:74.08 g/mol
- Merck Index:13,03841
- Boiling Pt:148 °C (1013 hPa)
- Melting Pt:–2 °C
- Density:0.9208 g/cm³ (20 °C)
- Flash Pt:–20 °C
- Storage Temperature:Ambient
- EINECS:203-721-0
- UN:1190
- ADR:3,II
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L



