Methyl hexanoate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:106-70-7
:
Formula:C₇H₁₄O₂
MDL Number:MFCD00009510
:130.19 g/mol
Boiling Pt:151 °C (1013 hPa)
Melting Pt:–63 °C
Density:0.88 g/cm³ (20 °C)
EINECS:203-425-1
ADR:3,III
:M-Clarity™ quality level = MQ300
UN:3272