Methyl hexanoate
Catalog # EHERC14196400
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:130.19 g/mol
Boiling Pt:151 °C (1013 hPa)
Melting Pt:–63 °C
Density:0.88 g/cm³ (20 °C)
EINECS:203-425-1
UN:3272
ADR:3,III
CAS Number:106-70-7
Formula:C₇H₁₄O₂
MDL Number:MFCD00009510