
Butyl butyrate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:109-21-7
:
Formula:C₈H₁₆O₂
MDL Number:MFCD00009450
Merck Index:14,1556
Boiling Pt:164…165 °C
Melting Pt:–92 °C
Density:0.871
Flash Pt:49 °C (120 °F)
UN:3272
ADR:3,III
:144.21 g/mol
EINECS:203-656-8
:GHS02
Storage Temperature:Ambient