Order Entry
Export
Orders LinkContactUsLinkComponent
 

Acetanilide 99+%

Acetanilide 99+%
Certificates
Acetanilide 99+%

5 Options Available Below

Product Details & Documents
Synonyms:  N-Phenylacetamide
  • CAS Number:103-84-4
  • Formula:C₈H₉NO
  • MDL Number:MFCD00008674
  • Molecular Weight:135.17 g/mol
  • Merck Index:13,00051
  • Boiling Pt:304 °C (760 mmHg)
  • Melting Pt:111…115 °C
  • Density:1,22 g/cm³ (20 °C)
  • Flash Pt:173 °C
  • Storage Temperature:Ambient
  • EINECS:203-150-7
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to beige to grey to light brown Powder or crystals or flakes
  • Infrared spectrum:
    Conforms
  • Melting point:
    111 to 115 °C
  • GC:
    ≥99.0 %
Product Family Options

Product Information