

Adipohydrazide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1071-93-8
:
Formula:C₆H₁₄N₄O₂
MDL Number:MFCD00007614
:174.2 g/mol
Boiling Pt:446 °C (1013 hPa)
Melting Pt:178…182 °C
Storage Temperature:Freezer
EINECS:213-999-5
UN:3077
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:178 to 182 °C
- Titration:≥97.5 %