Order Entry
Export
Orders LinkContactUsLinkComponent
ADA

ADA

Supplier:   Roth Carl
ADA
Supplier:  Roth Carl
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  2,2'-[(2-Amino-2-oxoethyl)imino]diacetic acid, N-(2-Acetamido)iminodiacetic acid, 2,2'-((2-Amino-2-oxoethyl)azanediyl)diacetic acid
  • CAS Number:26239-55-4
  • Formula:H₂NC(O)CH₂N(CH₂COOH)₂
  • MDL Number:MFCD00008031
  • Molecular Weight:190.16 g/mol
  • Merck Index:13,00148
  • Melting Pt:217…218 °C
  • Storage Temperature:Ambient
  • EINECS:247-530-0
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 33,20 €
    33,20 €
    1 * 50 g
       33,20 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 59,70 €
    59,70 €
    1 * 100 g
       59,70 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 231,00 €
    231,00 €
    1 * 500 g
       231,00 €