Formamide
Plastic bottle, 500 mL
Catalog # ROCH11814320001
Supplier: ROCHE
Jump to:
Product Details & Documents
Synonyms: Carbamaldehyde
- CAS Number:75-12-7
- Formula:HCONH₂
- MDL Number:MFCD00007941
- Molecular Weight:45.04 g/mol
- Merck Index:13,04261
- Boiling Pt:210 °C (1013 hPa)
- Melting Pt:2 °C
- Density:1.1334 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Flash Pt:79 °C
- Storage Temperature:Refrigerator
- EINECS:200-842-0
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:500 mL



