Formamide 99.5+%, extra pure
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Carbamaldehyde
- CAS Number:75-12-7
- Formula:HCONH₂
- MDL Number:MFCD00007941
- Molecular Weight:45.04 g/mol
- Merck Index:13,04261
- Boiling Pt:210 °C (760 mmHg)
- Melting Pt:2…3 °C
- Density:1.133 g/cm³
- Flash Pt:175 °C
- Storage Temperature:Ambient
- EINECS:200-842-0
3 Options Available Below
Product Details & Documents
Synonyms: Carbamaldehyde
- CAS Number:75-12-7
- Formula:HCONH₂
- MDL Number:MFCD00007941
- Molecular Weight:45.04 g/mol
- Merck Index:13,04261
- Boiling Pt:210 °C (760 mmHg)
- Melting Pt:2…3 °C
- Density:1.133 g/cm³
- Flash Pt:175 °C
- Storage Temperature:Ambient
- EINECS:200-842-0
Documents
ACRO18109
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4460 to 1.4480 (20°C, 589 nm)
- Water:≤0.1 % (Coulometric)
- GC:≥99.50 %
- Residue after evaporation:≤0.05 %



