N,N-Dimethylacetamide
Supplier: Roth Carl
Jump to:
About this item
Synonyms: Acetic acid dimethylamide, Dimethyl Acetamide, DMAC
- CAS Number:127-19-5
- Formula:CH₃CON(CH₃)₂
- MDL Number:MFCD00008686
- Molecular Weight:87.12 g/mol
- Merck Index:13,03254
- Boiling Pt:166.1 °C (1013 hPa)
- Melting Pt:–20 °C
- Density:0.937 g/cm³ (20 °C)
- Flash Pt:70 °C
- Storage Temperature:Ambient
- EINECS:204-826-4
- UN:2810
- ADR:6.1,III
2 Options Available Below
Product Details & Documents
Synonyms: Acetic acid dimethylamide, Dimethyl Acetamide, DMAC
- CAS Number:127-19-5
- Formula:CH₃CON(CH₃)₂
- MDL Number:MFCD00008686
- Molecular Weight:87.12 g/mol
- Merck Index:13,03254
- Boiling Pt:166.1 °C (1013 hPa)
- Melting Pt:–20 °C
- Density:0.937 g/cm³ (20 °C)
- Flash Pt:70 °C
- Storage Temperature:Ambient
- EINECS:204-826-4
- UN:2810
- ADR:6.1,III



