Order Entry
Export
Orders LinkContactUsLinkComponent
N,N-Dimethylacetamide ≥99%

N,N-Dimethylacetamide ≥99%

Health
Irritant
N,N-Dimethylacetamide ≥99%
Certificates
N,N-Dimethylacetamide ≥99%
N,N-Dimethylacetamide ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 127-19-5 Molecular Formula: C4H9NO Formula Weight: 87.12 Flash Point: 70°(158°F) Refractive Index: 1.4385 Odour: Ammonia-like Melting Point: -20° Boiling Point: 165-166° Beilstein Reference: 1737614 Merck Reference: 14,3227 MDL No.: MFCD00008686

Synonyms:  Acetic acid dimethylamide, Dimethyl Acetamide, DMAC
  • CAS Number:127-19-5
  • Formula:CH₃CON(CH₃)₂
  • MDL Number:MFCD00008686
  • Molecular Weight:87.12 g/mol
  • Merck Index:14,3227
  • Boiling Pt:165…166 °C
  • Melting Pt:–20 °C
  • Density:0.940
  • Flash Pt:70 °C (158 °F)
  • Storage Temperature:Ambient
  • EINECS:204-826-4
  • UN:2810
  • ADR:6.1,III
Product Family Options

Product Information

This product has been blocked by your organization. Please contact your purchasing department for more information