3,4-Hexanedione, Sigma-Aldrich®
Catalog # SIAL306932-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:114.14 g/mol
Boiling Pt:129 °C (1013 hPa)
Melting Pt:–10 °C
Density:0.9489 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:224-651-7
UN:1224
ADR:3,III
:M-Clarity™ quality level = MQ100
CAS Number:4437-51-8
Formula:C₆H₁₀O₂
MDL Number:MFCD00010237