Order Entry
Export
Orders LinkContactUsLinkComponent
Pyruvic acid

Pyruvic acid

Glass bottle for solids, 500 g
Catalog # MOLE28177601-500G
Corrosive
Supplier:   Molekula
Pyruvic acid
Catalog # MOLE28177601-500G
Supplier:  Molekula
CAS Number:  127-17-3
Pyruvic acid

Glass bottle for solids, 500 g

CAS Number:  127-17-3
Price
270,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  α-Ketopropionic acid, 2-Oxopropionic acid, 2-Ketopropionic acid, 2-Oxopropanoic acid
  • CAS Number:127-17-3
  • Formula:C₃H₄O₃
  • MDL Number:MFCD00002585
  • Molecular Weight:88.06 g/mol
  • Merck Index:12,08205
  • Boiling Pt:165 °C (1013 hPa)
  • Melting Pt:12 °C
  • Density:1.58 g/cm³ (20 °C)
  • Flash Pt:82 °C
  • Storage Temperature:Refrigerator
  • EINECS:204-824-3
  • UN:3265
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications