Pyruvic acid
100
Catalog # MEGAK-PYRUV
Supplier: MEGAZYME
Jump to:
Product Details & Documents
Synonyms: α-Ketopropionic acid, 2-Oxopropionic acid, 2-Ketopropionic acid, 2-Oxopropanoic acid
- CAS Number:127-17-3
- Formula:C₃H₄O₃
- MDL Number:MFCD00002585
- Molecular Weight:88.06 g/mol
- Merck Index:12,08205
- Boiling Pt:165 °C (1013 hPa)
- Melting Pt:12 °C
- Density:1.58 g/cm³ (20 °C)
- Flash Pt:82 °C
- Storage Temperature:Refrigerator
- EINECS:204-824-3
- UN:3265
- ADR:8,II
Specifications
Specifications
- Pk:100



