Order Entry
Export
Orders LinkContactUsLinkComponent
Diethyl pyrocarbonate

Diethyl pyrocarbonate

Irritant
Supplier:   Molekula
Diethyl pyrocarbonate
Supplier:  Molekula
Certificates
Diethyl pyrocarbonate

2 Options Available Below

Product Details & Documents
Synonyms:  DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC
  • CAS Number:1609-47-8
  • Formula:O(COOC₂H₅)₂
  • MDL Number:MFCD00009106
  • Molecular Weight:162.14 g/mol
  • Merck Index:13,08088
  • Boiling Pt:93 °C (1013 hPa)
  • Density:1.123 g/cm³ (20 °C)
  • Flash Pt:69 °C
  • Storage Temperature:Refrigerator
  • EINECS:216-542-8
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 150,00 €
    150,00 €
    1 * 25 g
       150,00 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 460,00 €
    460,00 €
    1 * 100 g
       460,00 €