Order Entry
Export
Orders LinkContactUsLinkComponent
 

Chloral hydrate

Toxic
Supplier:   Roth Carl
Chloral hydrate
Supplier:  Roth Carl
Certificates
Chloral hydrate

2 Options Available Below

Product Details & Documents
Synonyms:  2,2,2-Trichloroethane-1,1-diol
  • CAS Number:302-17-0
  • Formula:Cl₃CCH(OH)₂
  • MDL Number:MFCD00044479
  • Molecular Weight:165.4 g/mol
  • Merck Index:12,02113
  • Boiling Pt:97.5 °C (1013 hPa)
  • Melting Pt:52 °C
  • Density:1.901 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:206-117-5
  • ADR:6.1,III
  •  :Density: 1.901 g/cm³ (20 °C)
  • UN:2811
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 145,00 €
    145,00 €
    1 * 500 g
       145,00 €

  • Glass bottle for solids
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 608,00 €
    608,00 €
    1 * 2,5 kg
       608,00 €