1,2-Bis(2-chloroethoxy)ethane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:112-26-5
:
Formula:ClH₂CCH₂OCH₂CH₂OCH₂CH₂Cl
MDL Number:MFCD00000976
:187.07 g/mol
Boiling Pt:235 °C (1013 hPa)
Melting Pt:–31.5 °C
Density:1.1024 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-952-7
ADR:6.1,III
:M-Clarity™ quality level = MQ100
UN:2810