
1,1-Dichloro-2,2-diethoxyethane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:619-33-0
:
Formula:Cl₂HCCH(OCH₂CH₃)₂
MDL Number:MFCD00000849
:187.07 g/mol
Boiling Pt:152…154 °C (1013 hPa)
Density:1.4839 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:210-591-9
UN:1993
ADR:3,III
:M-Clarity™ quality level = MQ100
:GHS07