1,3-Dichloroacetone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:534-07-6
:
Formula:ClH₂CC(O)CH₂Cl
MDL Number:MFCD00000937
:126.97 g/mol
Merck Index:13,03077
Boiling Pt:173 °C (1013 hPa)
Melting Pt:39…42 °C
Density:1.432 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:208-585-6
UN:2649
ADR:6.1,II
:M-Clarity™ quality level = MQ100
Flash Pt:95 °C