Order Entry
Export
Orders LinkContactUsLinkComponent
Dichloromethane

Dichloromethane

Health
Irritant
Supplier:   Roth Carl
Dichloromethane
Supplier:  Roth Carl
Certificates
Dichloromethane

4 Options Available Below

Product Details & Documents
Synonyms:  Methylene chloride, DCM
  • CAS Number:75-09-2
  • Formula:CH₂Cl₂
  • MDL Number:MFCD00000881
  • Molecular Weight:84.93 g/mol
  • Merck Index:12,06140
  • Boiling Pt:39.8 °C (1013 hPa)
  • Melting Pt:–95 °C
  • Density:1.322 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-838-9
  • UN:1593
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 20,30 €
    20,30 €
    1 * 100 mL
       20,30 €

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 54,50 €
    54,50 €
    1 * 1 L
       54,50 €

  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 102,00 €
    102,00 €
    1 * 2,5 L
       102,00 €

  • 25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L - 508,00 €
    508,00 €
    1 * 25 L
       508,00 €