1,2-Epoxyhexane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1436-34-6
:
Formula:C₆H₁₂O
MDL Number:MFCD00005154
:100.16 g/mol
Boiling Pt:116 °C (1013 hPa)
Density:0.8456 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:215-864-6
UN:1993
ADR:3,II
:M-Clarity™ quality level = MQ100