Order Entry
Export
ContactUsLinkComponent
11-Bromo-1-undecanol
11-Bromo-1-undecanol
Catalog # MOLE21074243-25G
Supplier:  Molekula
CAS Number:  
11-Bromo-1-undecanol
Catalog # MOLE21074243-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :251.21 g/mol
Boiling Pt:350 °C (1013 hPa)
Boiling Pt:350 °C (1013 hPa)
Melting Pt:50 °C
Storage Temperature:Ambient
EINECS:216-554-3
CAS Number:1611-56-9
Formula:HO(CH₂)₁₀CH₂Br
MDL Number:MFCD00004752