
11-Bromo-1-undecanol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1611-56-9
:
Formula:HO(CH₂)₁₀CH₂Br
MDL Number:MFCD00004752
:251.21 g/mol
Boiling Pt:165…170 °C (1 mmHg)
Melting Pt:42…47 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:216-554-3
:GHS07
Specification Test Results
- Appearance:White to beige Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:42 to 47 °C
- GC:≥96.0 %