Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
10-Bromo-1-decanol
10-Bromo-1-decanol, Sigma-Aldrich®
10-Bromo-1-decanol, Sigma-Aldrich®
10-Bromo-1-decanol, Sigma-Aldrich®
SIAL310891-5G
:
Merck
:
10-Bromo-1-decanol, Sigma-Aldrich®
SIAL310891-5G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
237.18 g/mol
:
M-Clarity™ quality level = MQ100
:
258-572-4
:
GHS07
:
Ambient
:
53463-68-6
:
HO(CH₂)₉CH₂Br
:
MFCD00041681