


1,3-Dibromo-2-propanol 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:96-21-9
:
Formula:BrH₂CCH(OH)CH₂Br
MDL Number:MFCD00000216
:217.89 g/mol
Boiling Pt:89 °C (1013 hPa)
Density:2.13 g/cm³
Flash Pt:46 °C
EINECS:202-489-8
UN:1987
ADR:3,III
:GHS02
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow to light brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5500 to 1.5530 (20°C, 589 nm)
- GC:≥94 %