Order Entry
Export
Orders LinkContactUsLinkComponent
 

Bronopol, Sigma-Aldrich®

Corrosive
Irritant
Environment
Supplier:   Merck
Bronopol, Sigma-Aldrich®
Supplier:  Merck
Certificates
Bronopol, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
Synonyms:  2-Bromo-2-nitro-1,3-propanediol, 2-Bromo-2-nitropropane-1,3-diol
  • CAS Number:52-51-7
  • Formula:HOCH₂BrC(NO₂)CH₂OH
  • MDL Number:MFCD00007390
  • Molecular Weight:199.99 g/mol
  • Merck Index:13,01432
  • Boiling Pt:388 °C (1013 hPa)
  • Melting Pt:130…133 °C
  • Density:1.91 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-143-0
  • ADR:4.1,III
  •  :M-Clarity™ quality level = MQ200
  • UN:3241

 

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 36,50 €
    36,50 €
    1 * 25 g
       36,50 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 112,00 €
    112,00 €
    1 * 100 g
       112,00 €