Bronopol
Glass bottle for solids, 100 g
Catalog # MOLE65898637-100G
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 2-Bromo-2-nitro-1,3-propanediol, 2-Bromo-2-nitropropane-1,3-diol
- CAS Number:52-51-7
- Formula:HOCH₂BrC(NO₂)CH₂OH
- MDL Number:MFCD00007390
- Molecular Weight:199.99 g/mol
- Merck Index:13,01432
- Boiling Pt:388 °C (1013 hPa)
- Melting Pt:130…133 °C
- Density:1.91 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-143-0
- UN:3241
- ADR:4.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



