11-Bromoundecanoic acid, Sigma-Aldrich®
SIALB82804-25G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
:
:354 °C (1013 hPa)
:48…49 °C
:1.24 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:265.19 g/mol
:Ambient
:220-602-9
:2834-05-1
:BrH₂C(CH₂)₉COOH
:MFCD00002732