Order Entry
Export
ContactUsLinkComponent
11-Bromoundecanoic acid, Sigma-Aldrich®
11-Bromoundecanoic acid, Sigma-Aldrich®
Catalog # SIAL165816-250G
Supplier:  Merck
CAS Number:  
11-Bromoundecanoic acid, Sigma-Aldrich®
Catalog # SIAL165816-250G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
Boiling Pt:354 °C (1013 hPa)
Melting Pt:48…49 °C
Density:1.24 g/cm³ (20 °C)
 :265.19 g/mol
Storage Temperature:Ambient
EINECS:220-602-9
CAS Number:2834-05-1
Formula:BrH₂C(CH₂)₉COOH
MDL Number:MFCD00002732