1,8-Dibromooctane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:4549-32-0
:
Formula:BrH₂C(CH₂)₆CH₂Br
MDL Number:MFCD00000277
:272.02 g/mol
Boiling Pt:270 °C (1013 hPa)
Melting Pt:15 °C
Density:1.46 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:224-912-5