
1,4-Dibromopentane, Sigma-Aldrich®
Catalog # SIAL252077-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:229.94 g/mol
Boiling Pt:200 °C (1013 hPa)
Melting Pt:–34 °C
Density:1.687 g/cm³ (15 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:210-967-2
:GHS07
CAS Number:626-87-9
Formula:BrH₂C(CH₂)₂BrCHCH₃
MDL Number:MFCD00000153