1,2-Dibromopropane, Sigma-Aldrich®
SIAL140961-100G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
:
:201.89 g/mol
:12,08038
:140 °C (1013 hPa)
:–55 °C
:1.932 g/cm³ (20 °C)
:Ambient
:201-139-1
:1993
:3,III
:M-Clarity™ quality level = MQ100
:78-75-1
:BrH₂CBrCHCH₃
:MFCD00000148