1,7-Octadiyne, Sigma-Aldrich®
SIAL161292-10G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
:
:106.17 g/mol
:136 °C (1013 hPa)
:–32 °C
:0.8169 g/cm³ (21 °C)
:Refrigerator
:212-815-0
:3295
:3,III
:M-Clarity™ quality level = MQ100
:GHS02
:GHS07
:871-84-1
:C₈H₁₀
:MFCD00008580