Order Entry
Export
Orders LinkContactUsLinkComponent
1,7-Octadiyne 98%

1,7-Octadiyne 98%

Flammable
Irritant
1,7-Octadiyne 98%
Certificates
1,7-Octadiyne 98%

2 Options Available Below

Product Details & Documents
Synonyms:  Octa-1,7-diyne
  • CAS Number:871-84-1
  • Formula:C₈H₁₀
  • MDL Number:MFCD00008580
  • Molecular Weight:106.17 g/mol
  • Boiling Pt:136 °C (1013 hPa)
  • Melting Pt:–32 °C
  • Density:0.8 g/cm³
  • Flash Pt:23 °C
  • Storage Temperature:Refrigerator
  • EINECS:212-815-0
  • UN:1993
  • ADR:3,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4455 to 1.4485 (20°C, 589 nm)
  • GC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 88,70 €
    88,70 €
    1 * 10 g
       88,70 €

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 303,00 €
    303,00 €
    1 * 50 g
       303,00 €