Order Entry
Export
ContactUsLinkComponent
1,6-Heptadiyne 98%
1,6-Heptadiyne 98%
Certificates
undefined
1,6-Heptadiyne 98%

Specification Test Results

  • Appearance:
    Clear yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4400 to 1.4440 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

Related Information