

2,5-Dimethyl-2,4-hexadiene, Sigma-Aldrich®
Catalog # SIAL344311-250ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mL
- Seq:2590
Specifications
About this item
:
:110.2 g/mol
Boiling Pt:133 °C (1013 hPa)
Melting Pt:11 °C
Density:0.7625 g/cm³ (20 °C)
EINECS:212-115-5
UN:3295
ADR:3,III
:M-Clarity™ quality level = MQ100
:GHS02
:GHS07
CAS Number:764-13-6
Formula:C₈H₁₄
MDL Number:MFCD00008904