1,9-Decadiene ≥97%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 97% CAS No.: 1647-16-1 Molecular Formula: CH2=CH(CH2)6CH=CH2 Formula Weight: 138.25 Flash Point: 42°(107°F) Physical Form: Liquid Density: 0.75 Refractive Index: 1.4325 Boiling Point: 169° UN No.: 3295 MDL No.: MFCD00008670
Synonyms: Deca-1,9-diene
- CAS Number:1647-16-1
- Formula:C₁₀H₁₈
- MDL Number:MFCD00008670
- Molecular Weight:138.25 g/mol
- Boiling Pt:169 °C
- Melting Pt:–71.8 °C
- Density:0.75
- Flash Pt:42 °C (107 °F)
- Storage Temperature:Ambient
- EINECS:216-711-6
- UN:3295
- ADR:3,III
2 Options Available Below
Product Details & Documents
Purity: 97% CAS No.: 1647-16-1 Molecular Formula: CH2=CH(CH2)6CH=CH2 Formula Weight: 138.25 Flash Point: 42°(107°F) Physical Form: Liquid Density: 0.75 Refractive Index: 1.4325 Boiling Point: 169° UN No.: 3295 MDL No.: MFCD00008670
Synonyms: Deca-1,9-diene
- CAS Number:1647-16-1
- Formula:C₁₀H₁₈
- MDL Number:MFCD00008670
- Molecular Weight:138.25 g/mol
- Boiling Pt:169 °C
- Melting Pt:–71.8 °C
- Density:0.75
- Flash Pt:42 °C (107 °F)
- Storage Temperature:Ambient
- EINECS:216-711-6
- UN:3295
- ADR:3,III



