Order Entry
Export
ContactUsLinkComponent
1,7-Octadiene ≥98%, Sigma-Aldrich®
1,7-Octadiene ≥98%, Sigma-Aldrich®
Catalog # SIALO2501-100G-A
Supplier:  Merck
CAS Number:  
1,7-Octadiene ≥98%, Sigma-Aldrich®
Catalog # SIALO2501-100G-A
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Octa-1,7-diene
 :
 :110.2 g/mol
Boiling Pt:121 °C (1013 hPa)
Melting Pt:–100 °C
Density:0.7313 g/cm³ (20 °C)
Flash Pt:5 °C
Storage Temperature:Ambient
EINECS:223-054-9
UN:2309
ADR:3,II
 :M-Clarity™ quality level = MQ100
 :GHS02
 :GHS08
CAS Number:3710-30-3
Formula:C₈H₁₄
MDL Number:MFCD00008668