

1,4-Pentadiene 99%
:591-93-5
:
:C₅H₈
:MFCD00008665
:68.12 g/mol
:26 °C (760 mmHg)
:–148 °C
:0.65 g/cm³
:–20 °C
:Refrigerator
:209-736-9
:3295
:3,I
:GHS02
:GHS08
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3884 to 1.3904 (20°C, 589 nm)
- GC:≥98.5 %
- Specific optical rotation:+63° to +67° (20°C, 589 nm) (c=4, methanol)