Order Entry
Export
Orders LinkContactUsLinkComponent
Sorbic acid

Sorbic acid

Irritant
Supplier:   Roth Carl
Sorbic acid
Supplier:  Roth Carl
Certificates
Sorbic acid

3 Options Available Below

Product Details & Documents
Synonyms:  (E,E)-2,4-Hexadienoic acid, (2E,4E)-2,4-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, 2,4-Hexadienoic acid, (2E,4E)-Hexa-2,4-dienoic acid
  • CAS Number:110-44-1
  • Formula:CH₃CH=CHCH=CHCOOH
  • MDL Number:MFCD00002703
  • Molecular Weight:112.13 g/mol
  • Merck Index:13,08793
  • Boiling Pt:228 °C (1013 hPa)
  • Melting Pt:132…135 °C
  • Density:1.204 g/cm³ (20 °C)
  • Flash Pt:127 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:203-768-7
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 60,10 €
    60,10 €
    1 * 250 g
       60,10 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 116,00 €
    116,00 €
    1 * 500 g
       116,00 €

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 217,00 €
    217,00 €
    1 * 1 kg
       217,00 €