Sorbic acid ≥99%
Purity: 99% CAS No.: 110-44-1 Molecular Formula: C6H8O2 Formula Weight: 112.13 Flash Point: 127°(260°F) Melting Point: 132-135° Boiling Point: 228° dec. Beilstein Reference: 1741831 Merck Reference: 14,8721 MDL No.: MFCD00002703
: (E,E)-2,4-Hexadienoic acid, (2E,4E)-2,4-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, 2,4-Hexadienoic acid, (2E,4E)-Hexa-2,4-dienoic acid
- :110-44-1
- :CH₃CH=CHCH=CHCOOH
- :MFCD00002703
- :112.13 g/mol
- :14,8721
- :228 °C (dec.)
- :132…135 °C
- :1.205
- :127 °C (260 °F)
- :Ambient
- :203-768-7
Purity: 99% CAS No.: 110-44-1 Molecular Formula: C6H8O2 Formula Weight: 112.13 Flash Point: 127°(260°F) Melting Point: 132-135° Boiling Point: 228° dec. Beilstein Reference: 1741831 Merck Reference: 14,8721 MDL No.: MFCD00002703
: (E,E)-2,4-Hexadienoic acid, (2E,4E)-2,4-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, 2,4-Hexadienoic acid, (2E,4E)-Hexa-2,4-dienoic acid
- :110-44-1
- :CH₃CH=CHCH=CHCOOH
- :MFCD00002703
- :112.13 g/mol
- :14,8721
- :228 °C (dec.)
- :132…135 °C
- :1.205
- :127 °C (260 °F)
- :Ambient
- :203-768-7
Product Family Options
- Pack typePk



