Order Entry
Export
Orders LinkContactUsLinkComponent
Linoleic acid

Linoleic acid

Glass bottle for solids, 1 g
Catalog # ROTH5315.1
Supplier:   Roth Carl
Linoleic acid
Catalog # ROTH5315.1
Supplier:  Roth Carl
CAS Number:  60-33-3
Linoleic acid

Glass bottle for solids, 1 g

CAS Number:  60-33-3
Price
84,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid
  • CAS Number:60-33-3
  • Formula:C₁₈H₃₂O₂
  • MDL Number:MFCD00064241
  • Molecular Weight:280.45 g/mol
  • Merck Index:13,05527
  • Boiling Pt:360 °C (1013 hPa)
  • Melting Pt:–5 °C
  • Density:0.9 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-470-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications